missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Disposable Manual 55mL Reagent Reservoirs

Catalog No. p-7078490
Change view
Click to view available options
Packaging:
1 reservoir per bag
Bulk
No. per Pack:
100 reservoirs/Pk.
20 bags/Pk.
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging No. per Pack Sterility
12111086 1 reservoir per bag 20 bags/Pk. Sterile
12111087 Bulk 100 reservoirs/Pk. Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12111086 Supplier BIOTIX™ Supplier No. 63300121
Only null left

Designed with angled sides and V-shaped bottom.

  • Made from polystyrene
  • With graduation marks

Specifications

Color Translucent
Capacity (Metric) 55 mL
Material Polystyrene
Sterility Sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
No. per Pack 20 bags/Pk.
Packaging 1 reservoir per bag
Quantity 25/Pk.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less