missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIOTIX™ Disposable Manual 25mL Reagent Reservoirs
Description
- Made from polystyrene
- With graduation marks
Specifications
Specifications
| Color | Opaque-white |
| Capacity (Metric) | 25 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reservoir |
| For Use With (Application) | Filling Multi-channel Pipettors |
| Height (Metric) | 24.13 mm |
| Length (Metric) | 132.33 mm |
| No. per Pack | 20 |
| Packaging | Pk of 20 |
| Show More |