missing translation for 'onlineSavingsMsg'
Learn More

Simtuzumab (LOXL2) - Research Grade Biosimilar, ProSci

Catalog No. 16834471
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16834471 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16834471 Supplier ProSci Llc Supplier No. 105740100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chinese Hamster (CHO) Recombinant Monoclonal Antibody

Simtuzumab (LOXL2) Recombinant Monoclonal Antibody specifically detects Simtuzumab (LOXL2).

Specifications

Antigen Simtuzumab (LOXL2)
Applications Not Available
Classification Recombinant Monoclonal
Conjugate Unconjugated
Formulation PBS buffer pH7.5
Gene Alias AB0024,GS-6624
Host Species Chinese Hamster (CHO)
Purification Method >95%
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Not Available
Content And Storage Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for 1-2 weeks. Store at -20 °C for 12 months. Store at -80 °C for long term storage.
Product Type Antibody
Form Purified
Isotype IgG4 κ
Research Category Neuroscience
Show More Show Less