missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Disposable 55mL Reagent Reservoirs

Catalog No. 12111085
Change view
Click to view available options
Quantity:
25/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12111085 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12111085 Supplier BIOTIX™ Supplier No. 63300120
Get Extra Pipette Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed with angled sides and V-shaped bottom.

  • Made from polystyrene
  • With graduation marks

Specifications

Color White
Capacity (Metric) 55 mL
Material Polystyrene
Sterility Sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Packaging 50 reservoirs/Pk.
Quantity 25/Pk.
Volume (Metric) Well 55 mL
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less