missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Disposable 3-Well Divided 25mL Reagent Reservoirs

Designed with angled sides and V-shaped bottom.

$83.00 - $782.18

Specifications

Capacity (Metric) 25 mL
Material Polystyrene
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Autoclavable Autoclavable
View More Specs

Products 3
Catalog Number Mfr. No. Sterility Packaging Price Quantity  
Catalog Number Mfr. No. Sterility Packaging Price Quantity  
12-111-092
View Documents
BIOTIX™
63300127
Sterile 5 reservoirs/Bag N/A
Only null left
 
12-111-093
View Documents
BIOTIX™
63300128
Sterile 20 reservoirs/Pk. N/A
Only null left
 
12-111-094
View Documents
BIOTIX™
63300129
Non-sterile 100 reservoirs/Pk. N/A
Only null left
 
Description

Description

  • Made from polystyrene
Specifications

Specifications

25 mL
Reservoir
Autoclavable
Single-use
Polystyrene
Filling Multi-channel Pipettors
12
V-bottom
Videos
SDS
Documents

Documents