missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Disposable 3-Well Divided 25mL Reagent Reservoirs

Catalog No. 12111092
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
100 reservoirs/Pk.
20 reservoirs/Pk.
5 reservoirs/Bag
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Packaging
12111092 Sterile 5 reservoirs/Bag
12111093 Sterile 20 reservoirs/Pk.
12111094 Non-sterile 100 reservoirs/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12111092 Supplier BIOTIX™ Supplier No. 63300127
Get Extra Pipette Products with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed with angled sides and V-shaped bottom.

  • Made from polystyrene

Specifications

Color Opaque-white
Capacity (Metric) 25 mL
Material Polystyrene
Sterility Sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Packaging 5 reservoirs/Bag
Quantity 50/Pk.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less