missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIOTIX™ Disposable 100mL Reagent Reservoirs
Description
- Angled sides and a V-shaped bottom allow maximum recovery of reagents
- All reservoirs feature internal graduated volume lines and pour-spouts on the corners
- Made from polystyrene
Specifications
Specifications
| Color | White |
| Description | For automated platforms |
| Capacity (Metric) | 100 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reservoir |
| For Use With (Application) | Filling Multi-channel Pipettors |
| Packaging | 5 reservoirs per bag |
| Quantity | 200/Cs. |
| Autoclavable | Autoclavable |
| Show More |