missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ Disposable 100mL Reagent Reservoirs

Catalog No. 12111083
Change view
Click to view available options
Bottom Shape:
Flat
V-bottom
Quantity:
100/Cs.
20/Cs.
200/Cs.
50/Cs.
Packaging:
1 reservoir per bag
5 reservoirs per bag
Bulk
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bottom Shape Quantity Packaging
12111083 V-bottom 20/Cs. 1 reservoir per bag
12111082 V-bottom 50/Cs. 5 reservoirs per bag
12111084 Flat 100/Cs. Bulk
12111098 Flat 200/Cs. 5 reservoirs per bag
12111100 V-bottom 100/Cs. Bulk
12111099 V-bottom 100/Cs. 1 reservoir per bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12111083 Supplier BIOTIX™ Supplier No. 63300118
Only null left

Designed for use with multi-channel pipetting applications and can accommodate up to 12-channel pipettes.

  • Angled sides and a V-shaped bottom allow maximum recovery of reagents
  • All reservoirs feature internal graduated volume lines and pour-spouts on the corners
  • Made from polystyrene

Specifications

Color White
Description With angled sides and V-shaped bottom
Capacity (Metric) 100 mL
Material Polystyrene
Sterility Sterile
Type Reservoir
For Use With (Application) Filling Multi-channel Pipettors
Packaging 1 reservoir per bag
Quantity 20/Cs.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less