missing translation for 'onlineSavingsMsg'
Learn More

Disperse Yellow 3, ≥96.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013817
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013817 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013817 Supplier Merck Emd Millipore Supplier No. 1134425MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disperse Yellow 3 is an yellow azo dye which is majorly used in textile industry. It also has carcinogenic properties.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 2832-40-8
Physical Form Neat
Linear Formula C15H15N3O2
MDL Number MFCD00002377
Melting Point 268°C to 270°C
Synonym 4-(2-Hydroxy-5-methylphenylazo)acetanilide; N-[4-(2-Hydroxy-5-methylphenylazo)phenyl]acetamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 269.30
Percent Purity ≥96.0% (HPLC)
Grade Analytical Standard
Show More Show Less