missing translation for 'onlineSavingsMsg'
Learn More

Disodium Succinate Hexahydrate 98.0+%, TCI America™

Supplier:  TCI America S0105500G

 View more versions of this product

Catalog No. S0105500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Disodium Succinate Hexahydrate
White
MFCD00002790
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
270.142
CHEBI:63686
≥98.0% (T)
6106-21-4
C4H16Na2O10
500 g
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
disodium;butanedioate;hexahydrate
3083938
162.05
Crystalline Powder

Chemical Identifiers

6106-21-4
270.142
ZBTUYCUNQBRXOR-UHFFFAOYSA-L
3083938
disodium;butanedioate;hexahydrate
C4H16Na2O10
MFCD00002790
disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
CHEBI:63686
C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0911

RTECSNumber : WM7751000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions