missing translation for 'onlineSavingsMsg'
Learn More

Disodium Succinate Hexahydrate 98.0+%, TCI America™

Catalog No. p-7135811
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
S010525G 25 g
S0105500G 500 g
2 options

Catalog No. S010525G

Supplier: TCI America S010525G

Only null left

Chemical Identifiers

CAS 6106-21-4
Molecular Formula C4H16Na2O10
Molecular Weight (g/mol) 270.142
MDL Number MFCD00002790
InChI Key ZBTUYCUNQBRXOR-UHFFFAOYSA-L
Synonym disodium succinate hexahydrate, sodium succinate hexahydrate, unii-u16qod6c4e, butanedioic acid, disodium salt, hexahydrate, disodium butanedioate hexahydrate, u16qod6c4e, disodium hexahydrate succinate, sodium succinate dibasic hexahydrate, butanedioic acid disodium salt, succinic acid disodium salt hexahydrate
PubChem CID 3083938
ChEBI CHEBI:63686
IUPAC Name disodium;butanedioate;hexahydrate
SMILES C(CC(=O)[O-])C(=O)[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 162.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Succinate Hexahydrate
EINECSNumber (2)-0911
RTECSNumber WM7751000
TSCA Yes