missing translation for 'onlineSavingsMsg'
Learn More

Disodium Pamidronate Hydrate 97.0+%, TCI America™

Numéro de catalogue. p-7127997
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D3921100MG 100 mg
D39211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. D3921100MG Fournisseur TCI America Code fournisseur D3921100MG
Il en reste null

Identifiants chimiques

CAS 57248-88-1
Molecular Formula C3H9NNa2O7P2
Molecular Weight (g/mol) 279.03
MDL Number MFCD00867072,MFCD11045314,MFCD00168777
InChI Key CEYUIFJWVHOCPP-UHFFFAOYSA-L
Synonym Pamidronic Acid Disodium Salt, Disodium 3-Amino-1-hydroxypropane-1,1-diphosphonate
PubChem CID 131842120
IUPAC Name disodium hydrogen (3-amino-1-hydrogen phosphonato-1-hydroxypropyl)phosphonate
SMILES [Na+].[Na+].NCCC(O)(P(O)([O-])=O)P(O)([O-])=O

Spécifications

Color White
Quantity 100 mg
Formula Weight 279.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Pamidronate Hydrate
RTECSNumber SZ6525000
TSCA No
Recommended Storage Refrigerator