accessibility menu, dialog, popup

UserName

CAS RN 57248-88-1

NaNaH2NOHPOHOOPOHOO

CAS RN 57248-88-1

IUPAC Name: disodium hydrogen (3-amino-1-hydrogen phosphonato-1-hydroxypropyl)phosphonate
Synonyms: disodium hydrogen 3-amino-1-hydrogen phosphonato-1-hydroxypropylphosphonate
Molecular Weight (g/mol): 279.03
Molecular Formula: C3H9NNa2O7P2
InChi Key: CEYUIFJWVHOCPP-UHFFFAOYSA-L
SMILES: [Na+].[Na+].NCCC(O)(P(O)([O-])=O)P(O)([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results