missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Pamidronate Hydrate 97.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 1 g |
| Formula Weight | 279.03 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Disodium Pamidronate Hydrate |
Safety and Handling
| RTECSNumber | SZ6525000 |
| TSCA | No |
| Recommended Storage | Refrigerator |