missing translation for 'onlineSavingsMsg'
Learn More

Disodium Malonate, TCI America™

Catalog No. M0031500G
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0031500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0031500G Supplier TCI America Supplier No. M0031500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141-95-7
Molecular Formula C3H2Na2O4
Molecular Weight (g/mol) 148.025
MDL Number MFCD00002708
InChI Key PRWXGRGLHYDWPS-UHFFFAOYSA-L
Synonym sodium malonate, disodium malonate, propanedioic acid, disodium salt, malonic acid disodium salt, disodium propanedioate, malonic acid, sodium salt, unii-9qwe64m39h, propanedioic acid, sodium salt 1:2, malonic acid, disodium salt, dicarboxymethane disodium salt
PubChem CID 8865
ChEBI CHEBI:62983
IUPAC Name disodium;propanedioate
SMILES C(C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 148.02
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Malonate
EINECSNumber (9)-1371
RTECSNumber OO1750000
TSCA Yes