accessibility menu, dialog, popup

UserName

CAS RN 141-95-7

IUPAC Name: disodium propanedioate
Synonyms: sodium malonate propanedioic acid, sodium salt (1:2) sodium malonate dibasic propanedioic acid, disodium salt malonic acid, sodium salt malonic acid, disodium salt malonic acid disodium salt
Molecular Weight (g/mol): 148.03
Molecular Formula: C3H2Na2O4
InChi Key: PRWXGRGLHYDWPS-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CC([O-])=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)