missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Maleate 99.0+%, TCI America™
Supplier: TCI America M0014500G
Specifications
| Disodium Maleate | |
| White | |
| MFCD00013059 | |
| Maleic Acid Disodium Salt | |
| C(=CC(=O)O)C(=O)O.[Na].[Na] | |
| 162.052 | |
| 160.04 | |
| Crystalline Powder |
| 371-47-1 | |
| C4H4Na2O4 | |
| 500 g | |
| VXXVUHAXJHEYFH-UAIGNFCESA-N | |
| (Z)-but-2-enedioic acid;sodium | |
| 87068830 | |
| ≥99.0% (T) |
Chemical Identifiers
| 371-47-1 | |
| 162.052 | |
| VXXVUHAXJHEYFH-UAIGNFCESA-N | |
| 87068830 | |
| C(=CC(=O)O)C(=O)O.[Na].[Na] |
| C4H4Na2O4 | |
| MFCD00013059 | |
| Maleic Acid Disodium Salt | |
| (Z)-but-2-enedioic acid;sodium |
Safety and Handling
EINECSNumber : (2)-1102
RTECSNumber : ON2000000
TSCA : Yes