missing translation for 'onlineSavingsMsg'
Learn More

Disodium Maleate 99.0+%, TCI America™

Catalog No. p-7131927
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M001425G 25 g
M0014500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M001425G Supplier TCI America Supplier No. M001425G
Only null left

Chemical Identifiers

CAS 371-47-1
Molecular Formula C4H4Na2O4
Molecular Weight (g/mol) 162.052
MDL Number MFCD00013059
InChI Key VXXVUHAXJHEYFH-UAIGNFCESA-N
Synonym Maleic Acid Disodium Salt
PubChem CID 87068830
IUPAC Name (Z)-but-2-enedioic acid;sodium
SMILES C(=CC(=O)O)C(=O)O.[Na].[Na]

Specifications

Color White
Quantity 25 g
Formula Weight 160.04
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Maleate
EINECSNumber (2)-1102
RTECSNumber ON2000000
TSCA Yes