missing translation for 'onlineSavingsMsg'
Learn More

Disodium Glutarate 99.0+%, TCI America™

Catalog No. p-7130130
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G007025G 25 g
G0070500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G007025G Supplier TCI America Supplier No. G007025G
Only null left

Chemical Identifiers

CAS 13521-83-0
Molecular Formula C5H6Na2O4
Molecular Weight (g/mol) 176.08
MDL Number MFCD00070497
InChI Key ZUDYLZOBWIAUPC-UHFFFAOYSA-L
Synonym Glutaric Acid Disodium Salt
PubChem CID 161742
IUPAC Name disodium pentanedioate
SMILES [Na+].[Na+].[O-]C(=O)CCCC([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 176.08
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Glutarate