accessibility menu, dialog, popup

UserName

CAS RN 13521-83-0

IUPAC Name: disodium pentanedioate
Synonyms: disodium glutarate
Molecular Weight (g/mol): 176.08
Molecular Formula: C5H6Na2O4
InChi Key: ZUDYLZOBWIAUPC-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CCCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results