missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Fumarate 98.0+%, TCI America™
$52.19 - $171.49
Chemical Identifiers
| CAS | 17013-01-3 |
|---|---|
| Molecular Formula | C4H4Na2O4 |
| Molecular Weight (g/mol) | 162.052 |
| MDL Number | MFCD00064567 |
| InChI Key | VXXVUHAXJHEYFH-SEPHDYHBSA-N |
| Synonym | disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate |
| PubChem CID | 87110070 |
| IUPAC Name | (E)-but-2-enedioic acid;sodium |
| SMILES | C(=CC(=O)O)C(=O)O.[Na].[Na] |
Chemical Identifiers
| 17013-01-3 | |
| 162.052 | |
| VXXVUHAXJHEYFH-SEPHDYHBSA-N | |
| 87110070 | |
| C(=CC(=O)O)C(=O)O.[Na].[Na] |
| C4H4Na2O4 | |
| MFCD00064567 | |
| disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate | |
| (E)-but-2-enedioic acid;sodium |
Specifications
| 17013-01-3 | |
| C4H4Na2O4 | |
| 25 g | |
| VXXVUHAXJHEYFH-SEPHDYHBSA-N | |
| (E)-but-2-enedioic acid;sodium | |
| 87110070 | |
| ≥98.0% (T) | |
| Disodium Fumarate |
| White-Yellow | |
| MFCD00064567 | |
| disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate | |
| C(=CC(=O)O)C(=O)O.[Na].[Na] | |
| 162.052 | |
| 160.04 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1094
RTECSNumber : LT1830000
TSCA : No