accessibility menu, dialog, popup

UserName

NaNaOOOO(Z)

CAS RN 17013-01-3

IUPAC Name: disodium (2Z)-but-2-enedioate
Synonyms: disodium fumarate
Molecular Weight (g/mol): 160.04
Molecular Formula: C4H2Na2O4
InChi Key: MSJMDZAOKORVFC-UAIGNFCESA-L
SMILES: [Na+].[Na+].[O-]C(=O)\C=C/C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)