missing translation for 'onlineSavingsMsg'
Learn More

Disodium Etidronate Hydrate 98.0+%, TCI America™

Catalog No. p-7128130
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41595G 5 g
D415925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41595G Supplier TCI America Supplier No. D41595G
Only null left

Chemical Identifiers

CAS 7414-83-7
Molecular Formula C2H6Na2O7P2
Molecular Weight (g/mol) 249.99
MDL Number MFCD00152567
InChI Key GWBBVOVXJZATQQ-UHFFFAOYSA-L
Synonym etidronate disodium, disodium etidronate, didronel, sodium etidronate, sodium ethidronate, sodium ethydronate, disodium ethydronate, didronel r, disodium ethanol-1,1-diphosphonate, etidronsaeure dinatriumsalz
PubChem CID 23894
ChEBI CHEBI:4906
IUPAC Name disodium hydrogen (1-hydrogen phosphonato-1-hydroxyethyl)phosphonate
SMILES [Na+].[Na+].CC(O)(P(O)([O-])=O)P(O)([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 249.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Etidronate Hydrate
EINECSNumber (2)-2936
RTECSNumber SZ8562240
TSCA Yes
Recommended Storage Refrigerator