accessibility menu, dialog, popup

UserName

CAS RN 7414-83-7

IUPAC Name: disodium hydrogen (1-hydrogen phosphonato-1-hydroxyethyl)phosphonate
Synonyms: disodium etidronate
Molecular Weight (g/mol): 249.99
Molecular Formula: C2H6Na2O7P2
InChi Key: GWBBVOVXJZATQQ-UHFFFAOYSA-L
SMILES: [Na+].[Na+].CC(O)(P(O)([O-])=O)P(O)([O-])=O
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)