missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium EDTA, Dihydrate, FCC, 99-101%, Spectrum™ Chemical
E1001, 6381-92-6, C10H14N2Na2O8·2H2O
$360.69 - $9918.54
Chemical Identifiers
| CAS | 6381-92-6 |
|---|---|
| Molecular Formula | C10H18N2Na2O10 |
| Molecular Weight (g/mol) | 372.24 |
| MDL Number | MFCD00150037,MFCD00003541 |
| InChI Key | OVBJJZOQPCKUOR-UHFFFAOYSA-L |
| IUPAC Name | disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
| SMILES | O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602264
|
Spectrum Chemical Mfg Cor
E1001125GM |
125 g | Poly Bottle |
Each for $360.69
|
|
||||
|
18602265
|
Spectrum Chemical Mfg Cor
E1001500GM |
500 g | Poly Bottle |
Each for $891.03
|
|
||||
|
18602266
|
Spectrum Chemical Mfg Cor
E100125KG |
2.5 kg | Poly Pail |
Each for $3,119.69
|
|
||||
|
18602267
|
Spectrum Chemical Mfg Cor
E100112KG |
12 kg | Poly Pail |
Each for $4,740.89
|
|
||||
|
18602268
|
Spectrum Chemical Mfg Cor
E100145KGBL |
45 kg | Fiber Drum |
Each for $9,918.54
|
|
||||
Description
Spectrum™ Chemical Disodium EDTA, Dihydrate, FCC is used as a preservative to inhibit discoloration. Spectrum Chemical manufactured FCC products, including all FCC Disodium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 6381-92-6 | |
| 372.24 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
| C10H18N2Na2O10 | |
| MFCD00150037,MFCD00003541 | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate |
Specifications
| 6381-92-6 | |
| Poly Bottle | |
| MFCD00150037,MFCD00003541 | |
| OVBJJZOQPCKUOR-UHFFFAOYSA-L | |
| disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate | |
| 99 to 101% |
| 100% | |
| C10H18N2Na2O10 | |
| 125 g | |
| O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
| 372.24 | |
| FCC |