missing translation for 'onlineSavingsMsg'
Learn More

Disodium EDTA, Dihydrate, FCC, 99-101%, Spectrum™ Chemical

Numéro de catalogue. 18602265
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18602265 500 g Poly Bottle
18602264 125 g Poly Bottle
18602266 2.5 kg Poly Pail
18602267 12 kg Poly Pail
18602268 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 18602265 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur E1001500GM
Il en reste null

E1001, 6381-92-6, C10H14N2Na2O8·2H2O

Spectrum™ Chemical Disodium EDTA, Dihydrate, FCC is used as a preservative to inhibit discoloration. Spectrum Chemical manufactured FCC products, including all FCC Disodium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Identifiants chimiques

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Spécifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade FCC