missing translation for 'onlineSavingsMsg'
Learn More

Disodium Clodronate Tetrahydrate 98.0+%, TCI America™

Supplier:  TCI America D41605G

 View more versions of this product

Catalog No. D41605G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Disodium Clodronate Tetrahydrate
White
MFCD02940846
Clodronic Acid Disodium Salt
C(P(=O)(O)[O-])(P(=O)(O)[O-])(Cl)Cl.C(P(=O)(O)[O-])(P(=O)(O)[O-])(Cl)Cl.O.O.O.O.[Na+].[Na+]
603.777
288.85
Crystalline Powder
88416-50-6
C2H12Cl4Na2O16P4-2
5 g
MSZARZDVBJBTIF-UHFFFAOYSA-J
disodium;[dichloro-[hydroxy(oxido)phosphoryl]methyl]-hydroxyphosphinate;tetrahydrate
133640198
≥98.0% (T)

Chemical Identifiers

88416-50-6
603.777
MSZARZDVBJBTIF-UHFFFAOYSA-J
133640198
C(P(=O)(O)[O-])(P(=O)(O)[O-])(Cl)Cl.C(P(=O)(O)[O-])(P(=O)(O)[O-])(Cl)Cl.O.O.O.O.[Na+].[Na+]
C2H12Cl4Na2O16P4-2
MFCD02940846
Clodronic Acid Disodium Salt
disodium;[dichloro-[hydroxy(oxido)phosphoryl]methyl]-hydroxyphosphinate;tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions