missing translation for 'onlineSavingsMsg'
Learn More

Disodium Clodronate Tetrahydrate 98.0+%, TCI America™

Catalog No. D41601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41601G 1 g
D41605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41601G Supplier TCI America Supplier No. D41601G

Chemical Identifiers

CAS 88416-50-6
Molecular Formula C2H12Cl4Na2O16P4-2
Molecular Weight (g/mol) 603.777
MDL Number MFCD02940846
InChI Key MSZARZDVBJBTIF-UHFFFAOYSA-J
Synonym Clodronic Acid Disodium Salt
PubChem CID 133640198
IUPAC Name disodium;[dichloro-[hydroxy(oxido)phosphoryl]methyl]-hydroxyphosphinate;tetrahydrate
SMILES C(P(=O)(O)[O-])(P(=O)(O)[O-])(Cl)Cl.C(P(=O)(O)[O-])(P(=O)(O)[O-])(Cl)Cl.O.O.O.O.[Na+].[Na+]

Specifications

Color White
Quantity 1 g
Formula Weight 288.85
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Clodronate Tetrahydrate