missing translation for 'onlineSavingsMsg'
Learn More

Disodium Anthraquinone-2,6-disulfonate 98.0+%, TCI America™

Catalog No. A030825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A030825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A030825G Supplier TCI America Supplier No. A030825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 853-68-9
Molecular Formula C14H6Na2O8S2
Molecular Weight (g/mol) 412.29
MDL Number MFCD00001230
InChI Key PKOVWEHDVFYKHL-UHFFFAOYSA-L
Synonym anthraquinone-2,6-disulfonic acid disodium salt, disodium anthraquinone-2,6-disulfonate, 9,10-anthraquinone-2,6-disulfonic acid disodium salt, 2,6-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, disodium salt, disodium 9,10-dioxo-9,10-dihydroanthracene-2,6-disulfonate, sodium anthraquinone-2,6-disulfonate, 2,6-anthracenedisulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt 1:2, disodium 9,10-dioxoanthracene-2,6-disulfonate, 9,10-dihydro-9,10-dioxo-2,6-anthracenedisulfonic acid disodium salt, disodium 9,10-anthraquinone-2,6-disulfonate
PubChem CID 70070
IUPAC Name disodium 9,10-dioxo-9,10-dihydroanthracene-2,6-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=O)C3=CC(=CC=C3C(=O)C2=C1)S([O-])(=O)=O

Specifications

Quantity 25 g
Formula Weight 412.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Anthraquinone-2,6-disulfonate
EINECSNumber (4)-0725
TSCA Yes