accessibility menu, dialog, popup

UserName

NaNaOSOOOOSOOO

CAS RN 853-68-9

IUPAC Name: disodium 9,10-dioxo-9,10-dihydroanthracene-2,6-disulfonate
Synonyms: disodium 9,10-dioxoanthracene-2,6-disulfonate
Molecular Weight (g/mol): 412.29
Molecular Formula: C14H6Na2O8S2
InChi Key: PKOVWEHDVFYKHL-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2C(=O)C3=CC(=CC=C3C(=O)C2=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results