missing translation for 'onlineSavingsMsg'
Learn More

Disodium alpha-Glycerophosphate Hydrate 85.0+%, TCI America™

Catalog No. p-7130141
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00965G 5 g
G009625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G00965G Supplier TCI America Supplier No. G00965G

Chemical Identifiers

CAS 1555-56-2
Molecular Formula C3H7Na2O6P
Molecular Weight (g/mol) 216.036
MDL Number MFCD00035539
InChI Key GEKBIENFFVFKRG-UHFFFAOYSA-L
Synonym sodium glycerophosphate, disodium alpha-glycerophosphate, alpha-glycerophosphoric acid disodium salt, 1,2,3-propanetriol, mono dihydrogen phosphate , disodium salt, 1,2,3-propanetriol, 1-dihydrogen phosphate , disodium salt, dipotassium glyceryl phosphate, 1,2,3-propanetriol, 1-dihydrogen phosphate , sodium salt 1:2, 1,2,3-propanetriol, mono dihydrogen phosphate , sodium salt 1:2, 57-03-4 parent, sodium 1-glycerophosphate
PubChem CID 14754
IUPAC Name disodium;2,3-dihydroxypropyl phosphate
SMILES C(C(COP(=O)([O-])[O-])O)O.[Na+].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 216.04
Percent Purity ≥85.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium alpha-Glycerophosphate Hydrate
EINECSNumber (2)-1905&(2)-1990
TSCA Yes