accessibility menu, dialog, popup

UserName

NaNaOHHOOPOOO

CAS RN 1555-56-2

IUPAC Name: disodium 3-(phosphonatooxy)propane-1,2-diol
Synonyms: sodium glycerophosphate 甘油磷酸钠
Molecular Weight (g/mol): 216.04
Molecular Formula: C3H7Na2O6P
InChi Key: GEKBIENFFVFKRG-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OCC(O)COP([O-])([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results