missing translation for 'onlineSavingsMsg'
Learn More

Disodium Adipate 98.0+%, TCI America™

Catalog No. p-7116965
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A059225G 25 g
A0592500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A059225G Supplier TCI America Supplier No. A059225G
Only null left

Chemical Identifiers

CAS 7486-38-6
Molecular Formula C6H8Na2O4
Molecular Weight (g/mol) 190.106
MDL Number MFCD00070498
InChI Key KYKFCSHPTAVNJD-UHFFFAOYSA-L
Synonym Adipic Acid Disodium Salt
PubChem CID 24073
IUPAC Name disodium;hexanedioate
SMILES C(CCC(=O)[O-])CC(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 190.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Adipate
EINECSNumber (2)-0859
RTECSNumber AV1770000
TSCA Yes