accessibility menu, dialog, popup

UserName

CAS RN 7486-38-6

NaNaOOOO

CAS RN 7486-38-6

IUPAC Name: disodium hexanedioate
Synonyms: sodium adipate
Molecular Weight (g/mol): 190.11
Molecular Formula: C6H8Na2O4
InChi Key: KYKFCSHPTAVNJD-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CCCCC([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results