missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4-Amino-4'-nitrostilbene-2,2'-sulfonate 80.0+%, TCI America™

Catalog No. p-7117304
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
A119925G 25 g
A1199500G 500 g
2 options

Catalog No. A119925G

Supplier: TCI America A119925G

Only null left

Chemical Identifiers

CAS 6634-82-8
Molecular Formula C14H12N2NaO8S2+
Molecular Weight (g/mol) 423.366
MDL Number MFCD00143251
InChI Key DVSCXUQCFDWMOE-TYYBGVCCSA-N
PubChem CID 23669643
IUPAC Name sodium;5-amino-2-[(E)-2-(4-nitro-2-sulfophenyl)ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)[N+](=O)[O-])S(=O)(=O)O.[Na+]

Specifications

Quantity 25 g
Synonym 4-Amino-4′C-nitrostilbene-2,2′C-disulfonic Acid Disodium Salt
Formula Weight 444.34
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4-Amino-4′-nitrostilbene-2,2′-sulfonate