accessibility menu, dialog, popup

UserName

CAS RN 6634-82-8

IUPAC Name: sodium 5-amino-2-[2-(4-nitro-2-sulfophenyl)ethenyl]benzene-1-sulfonic acid
Synonyms: sodium 5-amino-2-[2-(4-nitro-2-sulfophenyl)ethenyl]benzenesulfonic acid
Molecular Weight (g/mol): 423.37
Molecular Formula: C14H12N2NaO8S2
InChi Key: DVSCXUQCFDWMOE-UHFFFAOYSA-N
SMILES: [Na+].NC1=CC(=C(C=CC2=C(C=C(C=C2)[N+]([O-])=O)S(O)(=O)=O)C=C1)S(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results