missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4-Amino-4'-nitrostilbene-2,2'-sulfonate 80.0+%, TCI America™

Catalog No. A1199500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1199500G 500 g
A119925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1199500G Supplier TCI America Supplier No. A1199500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6634-82-8
Molecular Formula C14H12N2NaO8S2+
Molecular Weight (g/mol) 423.366
MDL Number MFCD00143251
InChI Key DVSCXUQCFDWMOE-TYYBGVCCSA-N
PubChem CID 23669643
IUPAC Name sodium;5-amino-2-[(E)-2-(4-nitro-2-sulfophenyl)ethenyl]benzenesulfonic acid
SMILES C1=CC(=C(C=C1N)S(=O)(=O)O)C=CC2=C(C=C(C=C2)[N+](=O)[O-])S(=O)(=O)O.[Na+]

Specifications

Quantity 500 g
Synonym 4-Amino-4′-nitrostilbene-2,2′-disulfonic Acid Disodium Salt
Formula Weight 444.34
Percent Purity ≥80.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4-Amino-4′-nitrostilbene-2,2′-sulfonate