missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4,4'-Dinitrostilbene-2,2'-disulfonate 98.0+%, TCI America™

Catalog No. D0853500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0853500G 500 g
D085325G 25 g
D0853100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0853500G Supplier TCI America Supplier No. D0853500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3709-43-1
Molecular Formula C14H8N2Na2O10S2
Molecular Weight (g/mol) 474.32
MDL Number MFCD00024929
InChI Key SDCDTWFAOKXZHD-SEPHDYHBSA-L
PubChem CID 21116162
IUPAC Name disodium 5-nitro-2-[(1E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(\C=C\C2=CC=C(C=C2S([O-])(=O)=O)[N+]([O-])=O)C(=C1)S([O-])(=O)=O

Specifications

Quantity 500 g
Synonym 4,4′-Dinitrostilbene-2,2′-disulfonic Acid Disodium Salt
Formula Weight 474.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4,4′-Dinitrostilbene-2,2′-disulfonate