accessibility menu, dialog, popup

UserName

CAS RN 3709-43-1

IUPAC Name: disodium 5-nitro-2-[(1E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
Synonyms: disodium 5-nitro-2-[(1E)-2-(4-nitro-2-sulfonatophenyl)ethenyl]benzenesulfonate
Molecular Weight (g/mol): 474.32
Molecular Formula: C14H8N2Na2O10S2
InChi Key: SDCDTWFAOKXZHD-SEPHDYHBSA-L
SMILES: [Na+].[Na+].[O-][N+](=O)C1=CC=C(\C=C\C2=CC=C(C=C2S([O-])(=O)=O)[N+]([O-])=O)C(=C1)S([O-])(=O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results