missing translation for 'onlineSavingsMsg'
Learn More

Disodium 3-Hydroxy-2,7-naphthalenedisulfonate 85.0+%, TCI America™

Catalog No. N002925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N002925G 25 g
N0029500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N002925G Supplier TCI America Supplier No. N002925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-51-3
Molecular Formula C10H6Na2O7S2
Molecular Weight (g/mol) 348.251
MDL Number MFCD00065915
InChI Key VNEBWJSWMVTSHK-UHFFFAOYSA-L
Synonym Disodium 2-Naphthol-3,6-disulfonate, 3-Hydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, 2-Naphthol-3,6-disulfonic Acid Disodium Salt, R Acid Disodium Salt
PubChem CID 8673
IUPAC Name disodium;3-hydroxynaphthalene-2,7-disulfonate
SMILES C1=CC(=CC2=CC(=C(C=C21)O)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 348.25
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 3-Hydroxy-2,7-naphthalenedisulfonate
EINECSNumber (4)-0498
RTECSNumber QJ6477000
TSCA Yes