missing translation for 'onlineSavingsMsg'
Learn More

Disodium 3,3'-Dithiobis(1-propanesulfonate) 97.0+%, TCI America™

Catalog No. p-7128680
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D515925G 25 g
D5159100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D515925G Supplier TCI America Supplier No. D515925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27206-35-5
Molecular Formula C6H12Na2O6S4
Molecular Weight (g/mol) 354.376
MDL Number MFCD01104840
InChI Key WIYCQLLGDNXIBA-UHFFFAOYSA-L
Synonym 3,3′-Dithiobis(1-propanesulfonic Acid) Disodium Salt
PubChem CID 117948
IUPAC Name disodium;3-(3-sulfonatopropyldisulfanyl)propane-1-sulfonate
SMILES C(CSSCCCS(=O)(=O)[O-])CS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 354.38
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 3,3′-Dithiobis(1-propanesulfonate)
EINECSNumber (2)-1602
TSCA Yes