accessibility menu, dialog, popup

UserName

CAS RN 27206-35-5

NaNaOSOOSSSOOO

CAS RN 27206-35-5

IUPAC Name: disodium 3-[(3-sulfonatopropyl)disulfanyl]propane-1-sulfonate
Synonyms: disodium 3-[(3-sulfonatopropyl)disulfanyl]propane-1-sulfonate
Molecular Weight (g/mol): 354.38
Molecular Formula: C6H12Na2O6S4
InChi Key: WIYCQLLGDNXIBA-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)CCCSSCCCS([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results