missing translation for 'onlineSavingsMsg'
Learn More

Disodium 2-Oxoglutarate 98.0+%, TCI America™

Catalog No. p-7131738
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00075G 5 g
K000725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00075G Supplier TCI America Supplier No. K00075G

Chemical Identifiers

CAS 305-72-6
Molecular Formula C5H4Na2O5
Molecular Weight (g/mol) 190.062
MDL Number MFCD00043347
InChI Key YBGBJYVHJTVUSL-UHFFFAOYSA-L
Synonym disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
PubChem CID 31040
IUPAC Name disodium;2-oxopentanedioate
SMILES C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 190.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 2-Oxoglutarate
RTECSNumber SA0454700
TSCA Yes