accessibility menu, dialog, popup

UserName

CAS RN 305-72-6

IUPAC Name: disodium 2-oxopentanedioate
Synonyms: disodium α ketoglutarate
Molecular Weight (g/mol): 190.06
Molecular Formula: C5H4Na2O5
InChi Key: YBGBJYVHJTVUSL-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)CCC(=O)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)