missing translation for 'onlineSavingsMsg'
Learn More

Disodium 1,5-Naphthalenedisulfonate Hydrate 98.0+%, TCI America™

Catalog No. p-7133808
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N044125G 25 g
N0441500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N044125G Supplier TCI America Supplier No. N044125G
Only null left

Chemical Identifiers

CAS 1655-29-4
Molecular Formula C10H6Na2O6S2
Molecular Weight (g/mol) 332.252
MDL Number MFCD00064178
InChI Key YGSZNSDQUQYJCY-UHFFFAOYSA-L
Synonym disodium 1,5-naphthalenedisulfonate, sodium naphthalene-1,5-disulfonate, disodium naphthalene-1,5-disulfonate, 1,5-naphthalenedisulfonic acid, disodium salt, sodium 1,5-naphthalenedisulfonate, unii-28r71p6a5j, 1,5-naphthalenedisulfonic acid disodium salt, 1,5-naphthalenedisulfonic acid, sodium salt 1:2, 1,5-naphthalenedisulfonic acid disodium salt hydrate, 1,5-naphthalene disulfonic acid disudium salt
PubChem CID 74248
IUPAC Name disodium;naphthalene-1,5-disulfonate
SMILES C1=CC2=C(C=CC=C2S(=O)(=O)[O-])C(=C1)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 332.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1,5-Naphthalenedisulfonate Hydrate
EINECSNumber (4)-0468
TSCA Yes