missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-MCAX SPE Tube, Bed Wt. 300 mg

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

$235.90 - $295.84

Specifications

Column Capacity (Metric) 0.06 meq/g capacity
Column Phase DSC-MCAX
Column Type SPE
Bed Weight 300 mg
For Use With (Application) Food and Beverages
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity & Availability  
111010269
View Documents Promotion Details
MilliporeSigma™ Supelco™
52786U
6 mL 30/Pk.
Each for $235.90
Only null left
 
111009944
View Documents Promotion Details
MilliporeSigma™ Supelco™
52784U
3 mL 54/Pk.
Each for $295.84
Only null left
 
Description

Description

Retention Mechanism: Mixed-Mode Cation Exchange (reversed-phase and cation-exchange)Sample Matrix Compatibility: Aqueous Solutions (biological fluids, water) . Packed bed contains both octyl (C8) & benzene sulfonic acid (SCX) bondings. Dual retention mechanism broadens retention for a range of neutral, basic, acidic and zwitterionic compounds. Developed for superior selectivity/ sample clean up when isolating basic compounds from biological fluids. Greater ion-exchange capacity for isolating polar basic and zwitterionic compounds. Can be used to fractionate basic/ zwitterionic compounds from acidic and neutral compounds
Specifications

Specifications

0.06 meq/g capacity
SPE
Food and Beverages
Solid Phase Extraction Column
Polypropylene
480 m2/g
DSC-MCAX
300 mg
Discovery
Yes
70 Å
Videos
SDS
Documents

Documents

Discovery is a registered trademark of Sigma-Aldrich Co. LLC