missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-MCAX SPE Tube, Bed Wt. 300 mg

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Supplier:  MilliporeSigma™ Supelco™ 52784U

 View more versions of this product

Catalog No. 111009944


Only null left

Description

Description

Retention Mechanism: Mixed-Mode Cation Exchange (reversed-phase and cation-exchange)Sample Matrix Compatibility: Aqueous Solutions (biological fluids, water) . Packed bed contains both octyl (C8) & benzene sulfonic acid (SCX) bondings. Dual retention mechanism broadens retention for a range of neutral, basic, acidic and zwitterionic compounds. Developed for superior selectivity/ sample clean up when isolating basic compounds from biological fluids. Greater ion-exchange capacity for isolating polar basic and zwitterionic compounds. Can be used to fractionate basic/ zwitterionic compounds from acidic and neutral compounds
Specifications

Chemical Identifiers

0.06 meq/g capacity
SPE
Food and Beverages
Solid Phase Extraction Column
Polypropylene
480 m2/g
DSC-MCAX
300 mg
Discovery
Yes
70 Å

Specifications

0.06 meq/g capacity
SPE
Food and Beverages
Solid Phase Extraction Column
Yes
70 Å
480 m2/g
DSC-MCAX
300 mg
Discovery
3 mL
Polypropylene
54/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

Discovery is a registered trademark of Sigma-Aldrich Co. LLC