missing translation for 'onlineSavingsMsg'
Learn More

Disaccharides Kit, Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111016033
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016033 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016033 Supplier Merck Emd Millipore Supplier No. 47268U

Disaccharides Kit contains individually packaged standards in the quantities listed. The disaccharides are prepared, tested, and individually packaged using rigorous manufacturing procedures.

Specifications

Physical Form Neat
Quantity 1 Kit
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) Isomaltose (mixed anomers) 100 mg; alpha-Lactose 500 mg; Maltose (47288) 500 mg; Sucrose (47289) 500 mg