missing translation for 'onlineSavingsMsg'
Learn More

Direct Yellow 44, TCI America™

Catalog No. S014325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S014325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S014325G Supplier TCI America Supplier No. S014325G
Only null left

Chemical Identifiers

CAS 8005-52-5
Molecular Formula C27H20N6Na2O8S
Molecular Weight (g/mol) 634.531
MDL Number MFCD00059960
InChI Key LATMESDURBZWNH-UHFFFAOYSA-L
Synonym Direct Fast Yellow GC, Sirius Yellow GC
PubChem CID 57353166
IUPAC Name disodium;3-[[4-[[2-methoxy-4-[(3-sulfonatophenyl)diazenyl]phenyl]carbamoylamino]phenyl]hydrazinylidene]-6-oxocyclohexa-1,4-diene-1-carboxylate
SMILES COC1=C(C=CC(=C1)N=NC2=CC(=CC=C2)S(=O)(=O)[O-])NC(=O)NC3=CC=C(C=C3)NN=C4C=CC(=O)C(=C4)C(=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Formula Weight 634.53
Physical Form Crystalline Powder
Chemical Name or Material Direct Yellow 44
EINECSNumber (5)-1193
TSCA Yes