accessibility menu, dialog, popup

UserName

H3CONNSOOOHNOHNNHNOOONaNa(E/Z)(E/Z)

CAS RN 8005-52-5

IUPAC Name: disodium 3-(2-{4-[({2-methoxy-4-[2-(3-sulfonatophenyl)diazen-1-yl]phenyl}carbamoyl)amino]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate
Synonyms: disodium 3-(2-{4-[({2-methoxy-4-[2-(3-sulfonatophenyl)diazen-1-yl]phenyl}carbamoyl)amino]phenyl}hydrazin-1-ylidene)-6-oxocyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol): 634.53
Molecular Formula: C27H20N6Na2O8S
InChi Key: LATMESDURBZWNH-UHFFFAOYSA-L
SMILES: [Na+].[Na+].COC1=C(NC(=O)NC2=CC=C(NN=C3C=CC(=O)C(=C3)C([O-])=O)C=C2)C=CC(=C1)N=NC1=CC(=CC=C1)S([O-])(=O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results